siRNA Details
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020823.1 | NP_065874.1 | 17 | 2285 | ||
XM_005267074.3 | XP_005267131.1 | 17 | 2002 | |||
XM_011535999.1 | XP_011534301.1 | 17 | 2313 | |||
XM_011536000.1 | XP_011534302.1 | 15 | 2140 | |||
XM_011536001.2 | XP_011534303.1 | 17 | 2493 | |||
XM_011536002.2 | XP_011534304.1 | 17 | 2467 | |||
GenBank mRNA | AB037844.1 | - | 17 | 2287 | ||
BC033450.1 | - | 16 | 2008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
