siRNA Details
Gene Symbol: PRR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027031.1 | XP_016882520.1 | 7 | 4730 | ||
GenBank mRNA | AB033031.1 | - | 6 | 2462 | ||
BC034003.1 | - | 4 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
