siRNA Details
Gene Symbol: KIF26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015656.1 | NP_056471.1 | 15 | 6419 | ||
XM_011536641.2 | XP_011534943.1 | 12 | 5564 | |||
GenBank mRNA | AB033062.2 | - | 14 | 6293 | ||
AB290161.1 | - | 14 | 6389 | |||
AL117589.1 | - | 4 | 1573 | |||
BC009415.1 | - | 4 | 1607 | |||
BC025353.1 | - | 4 | 2565 | |||
BE540821.1 | - | 1 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
