siRNA Details
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080408.2 | NP_001073877.2 | 1, 2 | 725 | ||
GenBank mRNA | AK094068.1 | - | 1, 2 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
