siRNA Details
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080408.2 | NP_001073877.2 | 2 | 904 | ||
NM_001243007.1 | NP_001229936.1 | 3 | 1585 | |||
XM_005267543.3 | XP_005267600.1 | 6 | 2663 | |||
GenBank mRNA | AK094068.1 | - | 2 | 588 | ||
BC105720.1 | - | 3 | 1584 | |||
BC105927.1 | - | 3 | 1584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
