siRNA Details
Gene Symbol: MED29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317770.1 | NP_001304699.1 | 4 | 2562 | ||
NM_017592.2 | NP_060062.1 | 4 | 2632 | |||
NR_133915.1 | 3 | 2573 | ||||
GenBank mRNA | AK090877.1 | - | 3 | 2464 | ||
AK124629.1 | - | 5 | 2495 | |||
AL137304.1 | - | 4 | 2547 | |||
AY729650.1 | - | 4 | 2612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
