siRNA Details
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 11 | 1710 | ||
NM_001136030.2 | NP_001129502.1 | 11 | 1807 | |||
NM_001261844.1 | NP_001248773.1 | 10 | 1559 | |||
NM_014796.2 | NP_055611.1 | 9 | 1650 | |||
XM_005269247.2 | XP_005269304.1 | 10 | 1633 | |||
XM_006719715.3 | XP_006719778.1 | 13 | 2141 | |||
XM_011539035.2 | XP_011537337.1 | 13 | 2154 | |||
XM_011539036.1 | XP_011537338.1 | 12 | 1975 | |||
XM_011539037.2 | XP_011537339.1 | 11 | 6195 | |||
XM_017020262.1 | XP_016875751.1 | 13 | 3204 | |||
XM_017020263.1 | XP_016875752.1 | 13 | 3110 | |||
GenBank mRNA | AB018291.1 | - | 9 | 1848 | ||
AF161375.1 | - | 3 | 555 | |||
AK295026.1 | - | 9 | 1454 | |||
AK298405.1 | - | 9 | 1476 | |||
AK303223.1 | - | 11 | 1733 | |||
BC044578.1 | - | 9 | 1478 | 5275264 | ||
BC132732.1 | - | 11 | 1710 | 40146754 | ||
BC132734.1 | - | 11 | 1710 | 40146756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
