siRNA Details
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 9, 10 | 1582 | ||
NM_001136030.2 | NP_001129502.1 | 9, 10 | 1679 | |||
NM_001261844.1 | NP_001248773.1 | 8, 9 | 1431 | |||
NM_014796.2 | NP_055611.1 | 7, 8 | 1522 | |||
XM_005269247.2 | XP_005269304.1 | 8, 9 | 1505 | |||
XM_006719715.3 | XP_006719778.1 | 11, 12 | 2013 | |||
XM_011539035.2 | XP_011537337.1 | 11, 12 | 2026 | |||
XM_011539036.1 | XP_011537338.1 | 10, 11 | 1847 | |||
XM_011539037.2 | XP_011537339.1 | 9, 10 | 6067 | |||
XM_017020262.1 | XP_016875751.1 | 11, 12 | 3076 | |||
XM_017020263.1 | XP_016875752.1 | 11, 12 | 2982 | |||
GenBank mRNA | AB018291.1 | - | 7, 8 | 1720 | ||
AF161375.1 | - | 1, 2 | 428 | |||
AK295026.1 | - | 7, 8 | 1326 | |||
AK298405.1 | - | 7, 8 | 1348 | |||
AK303223.1 | - | 9, 10 | 1605 | |||
AK315972.1 | - | 8, 9 | 1385 | |||
BC044578.1 | - | 7, 8 | 1352 | 5275264 | ||
BC132732.1 | - | 9, 10 | 1582 | 40146754 | ||
BC132734.1 | - | 9, 10 | 1582 | 40146756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
