siRNA Details
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 6 | 864 | ||
NM_013328.3 | NP_037460.2 | 7 | 1086 | |||
GenBank mRNA | AF087859.1 | - | 7 | 1004 | ||
AF151351.1 | - | 7 | 883 | |||
AK001508.1 | - | 5 | 2465 | |||
AK291913.1 | - | 7 | 921 | |||
AL834217.1 | - | 3 | 819 | |||
BC014868.1 | - | 7 | 977 | 3919161 | ||
BC020553.1 | - | 7 | 1011 | 2901360 | ||
BU538765.1 | - | 6 | 739 | |||
KU178615.1 | - | 7 | 856 | |||
KU178616.1 | - | 7 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
