siRNA Details
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 6 | 936 | ||
NM_013328.3 | NP_037460.2 | 7 | 1158 | |||
GenBank mRNA | AF087859.1 | - | 7 | 1076 | ||
AF151351.1 | - | 7 | 955 | |||
AK001508.1 | - | 5 | 2537 | |||
AL834217.1 | - | 3 | 891 | |||
BC014868.1 | - | 7 | 1049 | 3919161 | ||
BC020553.1 | - | 7 | 1083 | 2901360 | ||
BU538765.1 | - | 6 | 812 | |||
KU178615.1 | - | 7 | 928 | |||
KU178616.1 | - | 7 | 752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
