siRNA Details
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 35 | 5766 | |||
GenBank mRNA | AB002391.2 | - | 32 | 5453 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
36 | 5766 | ||
AK095014.1 | - | 12 | 3049 | |||
CR749409.1 | - | 24 | 4471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
