siRNA Details
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 35 | 5463 | |||
GenBank mRNA | AB002391.2 | - | 32 | 5150 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
36 | 5462 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
31 | 5689 | ||
AK095014.1 | - | 12 | 2746 | |||
CR749409.1 | - | 24 | 4168 |
Gene Symbol: HERC2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036443.1 | 13 | 2351 | |||
GenBank mRNA | AF129930.1 | - | 1 | 279 | ||
AK056876.1 | - | 1 | 2171 | |||
AL832775.1 | - | 1 | 6714 | |||
BC047911.1 | - | 13 | 2351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
