siRNA Details
Gene Symbol: DDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164232.1 | NP_001157704.1 | 18 | 3023 | ||
NM_015214.2 | NP_056029.2 | 18 | 3200 | |||
XM_005273455.3 | XP_005273512.1 | 18 | 3384 | |||
XM_005273456.3 | XP_005273513.1 | 17 | 3258 | |||
XM_017013252.1 | XP_016868741.1 | 18 | 3157 | |||
XM_017013254.1 | XP_016868743.1 | 17 | 2937 | |||
XM_017013255.1 | XP_016868744.1 | 16 | 2946 | |||
GenBank mRNA | AB018268.1 | - | 15 | 2410 | ||
AK023218.1 | - | 14 | 2229 | |||
AK026961.1 | - | 3 | 821 | |||
AK056525.1 | - | 18 | 3102 | |||
AK125904.1 | - | 18 | 3023 | |||
AK127040.1 | - | 9 | 2551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
