siRNA Details
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 22, 23 | 2796 | ||
NM_014963.2 | NP_055778.2 | 25, 26 | 3128 | |||
XM_011527804.2 | XP_011526106.1 | 25, 26 | 3101 | |||
GenBank mRNA | AB023180.1 | - | 25, 26 | 3105 | ||
AK074102.1 | - | 21, 22 | 2588 | |||
AK125139.1 | - | 22, 23 | 2743 | |||
AK292407.1 | - | 25, 26 | 3128 | |||
BC106021.1 | - | 2, 3 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
