siRNA Details
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014936.4 | NP_055751.1 | 2 | 940 | ||
GenBank mRNA | AB020686.1 | - | 1 | 729 | ||
AK291276.1 | - | 2 | 785 | |||
BC018054.1 | - | 2 | 910 | 4797659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
