siRNA Details
Gene Symbol: CNPY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103176.1 | NP_001096646.1 | 4 | 1831 | ||
GenBank mRNA | AK094445.1 | - | 4 | 1849 | ||
BC105732.1 | - | 4 | 1832 | |||
BC105733.1 | - | 4 | 1832 | |||
BC105734.1 | - | 4 | 1832 | |||
BC107598.1 | - | 4 | 1832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
