siRNA Details
Gene Symbol: VARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167733.2 | NP_001161205.1 | 2 | 196 | ||
NM_001167734.1 | NP_001161206.1 | 3 | 431 | |||
NM_020442.5 | NP_065175.4 | 3 | 424 | |||
GenBank mRNA | AB067472.1 | - | 2 | 892 | ||
AK000511.1 | - | 3 | 311 | |||
AK293780.1 | - | 2 | 185 | |||
AK294451.1 | - | 3 | 368 | |||
AK303415.1 | - | 3 | 413 | |||
AK304807.1 | - | 3 | 431 | |||
BC112054.1 | - | 2 | 286 | 8327522 | ||
BC113605.1 | - | 2 | 286 | 8322657 | ||
BC143534.1 | - | 2 | 285 | |||
BC143535.1 | - | 2 | 285 | |||
BC143536.1 | - | 2 | 285 | 9052045 | ||
DC307585.1 | - | 3 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
