siRNA Details
Gene Symbol: VARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167733.2 | NP_001161205.1 | 10 | 937 | ||
NM_001167734.1 | NP_001161206.1 | 11 | 1172 | |||
NM_020442.5 | NP_065175.4 | 11 | 1165 | |||
GenBank mRNA | AB067472.1 | - | 10 | 1633 | ||
AK125069.1 | - | 1 | 79 | |||
AK293780.1 | - | 10 | 926 | |||
AK303415.1 | - | 11 | 1154 | |||
AK304807.1 | - | 11 | 1172 | |||
BC009355.2 | - | 1 | 13 | |||
BC112054.1 | - | 10 | 1027 | 8327522 | ||
BC113605.1 | - | 10 | 1027 | 8322657 | ||
BC143534.1 | - | 10 | 1026 | |||
BC143535.1 | - | 10 | 1026 | |||
BC143536.1 | - | 10 | 1026 | 9052045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
