siRNA Details
Gene Symbol: VARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167733.2 | NP_001161205.1 | 19 | 1757 | ||
NM_001167734.1 | NP_001161206.1 | 20 | 1992 | |||
NM_020442.5 | NP_065175.4 | 20 | 1985 | |||
GenBank mRNA | AB067472.1 | - | 19 | 2453 | ||
AK000511.1 | - | 19 | 1662 | |||
AK025618.1 | - | 8 | 577 | |||
AK125069.1 | - | 9 | 1740 | |||
AK293780.1 | - | 19 | 1746 | |||
AK303415.1 | - | 20 | 1974 | |||
AK304807.1 | - | 20 | 1992 | |||
AK315733.1 | - | 9 | 743 | |||
BC008844.2 | - | 3 | 180 | |||
BC009355.2 | - | 10 | 833 | |||
BC063427.1 | - | 8 | 608 | |||
BC073838.1 | - | 8 | 603 | |||
BC112054.1 | - | 19 | 1847 | 8327522 | ||
BC113605.1 | - | 19 | 1847 | 8322657 | ||
BC143534.1 | - | 18 | 1775 | |||
BC143535.1 | - | 19 | 1846 | |||
BC143536.1 | - | 19 | 1840 | 9052045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
