siRNA Details
Gene Symbol: GRAMD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286563.1 | NP_001273492.1 | 21 | 3113 | ||
NM_001286564.1 | NP_001273493.1 | 20 | 2822 | |||
NM_020716.2 | NP_065767.1 | 20 | 3092 | |||
XM_011542933.1 | XP_011541235.1 | 18 | 3234 | |||
XM_017018036.1 | XP_016873525.1 | 20 | 3532 | |||
XM_017018037.1 | XP_016873526.1 | 21 | 3831 | |||
XM_017018038.1 | XP_016873527.1 | 22 | 3077 | |||
XM_017018041.1 | XP_016873530.1 | 21 | 3135 | |||
XM_017018042.1 | XP_016873531.1 | 20 | 3123 | |||
XM_017018043.1 | XP_016873532.1 | 19 | 2756 | |||
XM_017018044.1 | XP_016873533.1 | 19 | 3080 | |||
XM_017018047.1 | XP_016873536.1 | 21 | 3578 | |||
XM_017018048.1 | XP_016873537.1 | 20 | 3534 | |||
XM_017018049.1 | XP_016873538.1 | 20 | 3563 | |||
XM_017018050.1 | XP_016873539.1 | 19 | 2839 | |||
XM_017018051.1 | XP_016873540.1 | 18 | 2455 | |||
GenBank mRNA | AB033027.1 | - | 20 | 2755 | ||
AY358924.1 | - | 9 | 2333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
