siRNA Details
Gene Symbol: SV2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297716.1 | NP_001284645.1 | 9, 10 | 1699 | ||
NM_014979.3 | NP_055794.3 | 9, 10 | 1699 | |||
XM_011543281.2 | XP_011541583.1 | 9, 10 | 2066 | |||
XM_011543282.2 | XP_011541584.2 | 9, 10 | 2626 | |||
XM_017009244.1 | XP_016864733.1 | 5, 6 | 617 | |||
GenBank mRNA | AB028977.1 | - | 7, 8 | 760 | ||
AK094917.1 | - | 9, 10 | 1700 | |||
BC100824.1 | - | 9, 10 | 1635 | 40001296 | ||
BC100825.1 | - | 9, 10 | 1633 | 40001297 | ||
BC100826.1 | - | 9, 10 | 1633 | 40001302 | ||
BC100827.1 | - | 9, 10 | 1633 | 40001303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
