siRNA Details
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 26 | 4443 | ||
XM_011523768.2 | XP_011522070.1 | 25 | 4374 | |||
GenBank mRNA | AB014564.1 | - | 22 | 3832 | ||
AK023003.1 | - | 18 | 3292 | |||
AK074252.1 | - | 11 | 1637 | |||
AK093541.1 | - | 1 | 1471 | |||
AK125717.1 | - | 26 | 4462 | |||
BC004266.1 | - | 8 | 1393 | |||
BC033614.1 | - | 8 | 1259 | |||
BC063786.1 | - | 20 | 3525 | |||
BX648864.1 | - | 4 | 2092 | |||
CR627449.1 | - | 24 | 4181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
