siRNA Details
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 11 | 1950 | ||
NM_001286762.1 | NP_001273691.1 | 10 | 1856 | |||
NM_001286763.1 | NP_001273692.1 | 11 | 1585 | |||
NM_001286764.1 | NP_001273693.1 | 10 | 1469 | |||
NM_001286765.1 | NP_001273694.1 | 10 | 1315 | |||
NM_001286766.1 | NP_001273695.1 | 9 | 1199 | |||
NM_025113.3 | NP_079389.2 | 11 | 1972 | |||
GenBank mRNA | AK025215.1 | - | 10 | 1605 | ||
AK093073.1 | - | 11 | 1920 | |||
AK125750.1 | - | 1 | 693 | |||
AK125950.1 | - | 11 | 1957 | |||
AK293034.1 | - | 10 | 1221 | |||
AK300183.1 | - | 10 | 1019 | |||
AK307854.1 | - | 9 | 942 | |||
AK310264.1 | - | 13 | 2060 | |||
BC004495.1 | - | 7 | 669 | |||
BC032311.1 | - | 9 | 2999 | |||
BC043488.1 | - | 11 | 1318 | 5205680 | ||
BX641126.1 | - | 9 | 1006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
