siRNA Details
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 8, 9 | 1709 | ||
NM_001286762.1 | NP_001273691.1 | 7, 8 | 1615 | |||
NM_001286763.1 | NP_001273692.1 | 8, 9 | 1344 | |||
NM_001286764.1 | NP_001273693.1 | 7, 8 | 1228 | |||
NM_001286765.1 | NP_001273694.1 | 7, 8 | 1074 | |||
NM_001286766.1 | NP_001273695.1 | 6, 7 | 958 | |||
NM_025113.3 | NP_079389.2 | 8, 9 | 1731 | |||
GenBank mRNA | AK025215.1 | - | 7, 8 | 1364 | ||
AK093073.1 | - | 8, 9 | 1679 | |||
AK125950.1 | - | 8, 9 | 1716 | |||
AK293034.1 | - | 7, 8 | 980 | |||
AK300183.1 | - | 7, 8 | 778 | |||
AK307854.1 | - | 6, 7 | 701 | |||
AK310264.1 | - | 9, 10 | 1718 | |||
BC004495.1 | - | 4, 5 | 428 | |||
BC032311.1 | - | 6, 7 | 1235 | |||
BC043488.1 | - | 8, 9 | 1077 | 5205680 | ||
BX641126.1 | - | 6, 7 | 765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
