siRNA Details
Gene Symbol: ATP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014616.2 | NP_055431.1 | 2 | 369 | ||
XM_005247241.2 | XP_005247298.1 | 2 | 368 | |||
XM_011512593.2 | XP_011510895.1 | 2 | 368 | |||
XM_011512594.2 | XP_011510896.1 | 2 | 368 | |||
XM_017006009.1 | XP_016861498.1 | 1 | 63 | |||
GenBank mRNA | AF156548.1 | - | 2 | 278 | ||
AK296750.1 | - | 2 | 368 | |||
BC042180.1 | - | 2 | 369 | |||
BI861412.1 | - | 2 | 369 | |||
DR000074.1 | - | 2 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
