siRNA Details
Gene Symbol: ATP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014616.2 | NP_055431.1 | 7 | 853 | ||
XM_005247241.2 | XP_005247298.1 | 7 | 852 | |||
XM_005247243.2 | XP_005247300.1 | 5 | 444 | |||
XM_011512593.2 | XP_011510895.1 | 7 | 852 | |||
XM_011512594.2 | XP_011510896.1 | 7 | 852 | |||
XM_011512597.2 | XP_011510899.1 | 5 | 419 | |||
XM_017006008.1 | XP_016861497.1 | 5 | 421 | |||
XM_017006009.1 | XP_016861498.1 | 6 | 547 | |||
GenBank mRNA | AF156548.1 | - | 7 | 762 | ||
BX489704.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
