siRNA Details
Gene Symbol: IQSEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111125.2 | NP_001104595.1 | 3 | 1167 | ||
NM_015075.1 | NP_055890.1 | 3 | 444 | |||
XM_006724579.2 | XP_006724642.1 | 4 | 1289 | |||
XM_006724580.3 | XP_006724643.1 | 3 | 457 | |||
XM_006724581.3 | XP_006724644.1 | 4 | 1289 | |||
XM_006724582.3 | XP_006724645.1 | 4 | 1289 | |||
XM_006724583.3 | XP_006724646.1 | 4 | 1289 | |||
XM_006724584.2 | XP_006724647.1 | 4 | 1289 | |||
XM_011530773.2 | XP_011529075.1 | 2 | 361 | |||
XM_011530774.2 | XP_011529076.1 | 4 | 1289 | |||
XM_011530776.1 | XP_011529078.1 | 4 | 1289 | |||
XM_011530777.1 | XP_011529079.1 | 4 | 1289 | |||
XM_017029359.1 | XP_016884848.1 | 2 | 1163 | |||
XM_017029360.1 | XP_016884849.1 | 3 | 575 | |||
GenBank mRNA | AB011094.1 | - | 3 | 1167 | ||
AK095232.1 | - | 3 | 444 | |||
DA433389.1 | - | 3 | 444 | |||
FJ154890.1 | - | 2 | 346 | |||
FJ154891.1 | - | 3 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
