siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 8 | 1143 | ||
NM_014985.3 | NP_055800.2 | 8 | 1143 | |||
XM_006720437.3 | XP_006720500.1 | 8 | 1328 | |||
XM_011521373.2 | XP_011519675.1 | 8 | 1896 | |||
XM_011521374.2 | XP_011519676.1 | 8 | 1895 | |||
XM_011521375.2 | XP_011519677.1 | 8 | 1895 | |||
XM_011521378.2 | XP_011519680.1 | 8 | 1895 | |||
XM_011521379.2 | XP_011519681.1 | 8 | 1894 | |||
XM_017022016.1 | XP_016877505.1 | 8 | 1898 | |||
GenBank mRNA | AB020719.1 | - | 7 | 748 | ||
AK303805.1 | - | 8 | 1066 | |||
BC069186.1 | - | 8 | 1039 | |||
BC107721.1 | - | 8 | 1147 | |||
BC117182.1 | - | 8 | 984 | 40125733 | ||
BX648822.1 | - | 8 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
