siRNA Details
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170738.1 | NP_001164209.1 | 14 | 4250 | ||
XM_011520961.1 | XP_011519263.1 | 13 | 3753 | |||
GenBank mRNA | AB029033.1 | - | 11 | 2925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
