siRNA Details
Gene Symbol: SSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198455.2 | NP_940857.2 | 93, 94 | 14251 | ||
GenBank mRNA | AK093431.1 | - | 4, 5 | 532 | ||
AK123170.1 | - | 9, 10 | 1264 | |||
AK124542.1 | - | 3, 4 | 1133 | |||
AY927598.1 | - | 8, 9 | 1058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
