siRNA Details
Gene Symbol: ADGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024694.3 | NP_078970.3 | 15 | 1847 | ||
XM_006715566.3 | XP_006715629.1 | 15 | 1882 | |||
XM_011536125.2 | XP_011534427.1 | 15 | 1833 | |||
XM_011536126.1 | XP_011534428.1 | 15 | 1882 | |||
XM_011536127.2 | XP_011534429.1 | 15 | 1882 | |||
XM_011536128.2 | XP_011534430.1 | 15 | 1882 | |||
XM_011536130.2 | XP_011534432.1 | 16 | 1781 | |||
XM_011536131.2 | XP_011534433.1 | 9 | 1055 | |||
XM_011536132.2 | XP_011534434.1 | 7 | 643 | |||
XM_011536134.2 | XP_011534436.1 | 12 | 1581 | |||
XM_017011314.1 | XP_016866803.1 | 15 | 1882 | |||
XM_017011315.1 | XP_016866804.1 | 15 | 1882 | |||
GenBank mRNA | AK097570.1 | - | 15 | 1847 | ||
AL832192.1 | - | 12 | 1583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
