siRNA Details
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512262.2 | XP_011510564.1 | 7, 8 | 906 | ||
XM_011512265.2 | XP_011510567.1 | 5, 6 | 385 | |||
XM_017005474.1 | XP_016860963.1 | 7, 8 | 906 | |||
XM_017005475.1 | XP_016860964.1 | 7, 8 | 906 | |||
XM_017005476.1 | XP_016860965.1 | 8, 9 | 936 | |||
XM_017005477.1 | XP_016860966.1 | 8, 9 | 2531 | |||
XM_017005478.1 | XP_016860967.1 | 8, 9 | 662 | |||
GenBank mRNA | AK092780.1 | - | 2, 3 | 255 | ||
BC041437.1 | - | 8, 9 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
