siRNA Details
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512262.2 | XP_011510564.1 | 1 | 39 | ||
XM_017005474.1 | XP_016860963.1 | 1 | 39 | |||
XM_017005475.1 | XP_016860964.1 | 1 | 39 | |||
GenBank mRNA | BC041437.1 | - | 1 | 42 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
