siRNA Details
Gene Symbol: CEP128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152446.3 | NP_689659.2 | 5 | 612 | ||
XM_006720056.3 | XP_006720119.1 | 6 | 793 | |||
XM_011536490.2 | XP_011534792.1 | 6 | 793 | |||
XM_011536491.2 | XP_011534793.1 | 6 | 793 | |||
XM_011536492.2 | XP_011534794.1 | 6 | 634 | |||
XM_011536493.2 | XP_011534795.1 | 6 | 793 | |||
XM_011536495.2 | XP_011534797.1 | 6 | 793 | |||
XM_017021042.1 | XP_016876531.1 | 7 | 911 | |||
GenBank mRNA | BC036939.1 | - | 1 | 85 | ||
BC045834.1 | - | 6 | 719 | |||
BC150610.1 | - | 4 | 470 | 9056980 | ||
BX248746.1 | - | 7 | 773 | |||
HQ215523.1 | - | 6 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
