siRNA Details
Gene Symbol: COL4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000092.4 | NP_000083.3 | 48 | 5622 | ||
XM_005246281.3 | XP_005246338.1 | 48 | 5156 | |||
XM_005246282.3 | XP_005246339.1 | 42 | 4739 | |||
XM_006712246.3 | XP_006712309.1 | 47 | 5105 | |||
XM_011510557.2 | XP_011508859.1 | 49 | 5213 | |||
XM_011510558.2 | XP_011508860.1 | 47 | 5186 | |||
XM_011510572.2 | XP_011508874.1 | 27 | 3519 | |||
XM_017003297.1 | XP_016858786.1 | 48 | 5177 | |||
GenBank mRNA | X81053.1 | - | 48 | 5176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
