siRNA Details
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 2 | 131 | ||
XM_011537009.1 | XP_011535311.1 | 2 | 223 | |||
XM_017021499.1 | XP_016876988.1 | 2 | 267 | |||
XM_017021500.1 | XP_016876989.1 | 2 | 223 | |||
XM_017021501.1 | XP_016876990.1 | 2 | 267 | |||
GenBank mRNA | AB033026.1 | - | 2 | 107 | ||
AK307428.1 | - | 2 | 223 | |||
BC146787.1 | - | 2 | 107 | 9007246 | ||
DA056171.1 | - | 2 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
