siRNA Details
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 28 | 4001 | ||
XM_011537009.1 | XP_011535311.1 | 28 | 4177 | |||
XM_011537015.2 | XP_011535317.1 | 21 | 4232 | |||
XM_017021499.1 | XP_016876988.1 | 27 | 4158 | |||
XM_017021500.1 | XP_016876989.1 | 28 | 4093 | |||
XM_017021501.1 | XP_016876990.1 | 27 | 4074 | |||
XM_017021502.1 | XP_016876991.1 | 21 | 3954 | |||
XM_017021503.1 | XP_016876992.1 | 21 | 3314 | |||
XM_017021504.1 | XP_016876993.1 | 23 | 3456 | |||
GenBank mRNA | AB033026.1 | - | 28 | 3977 | ||
AK124787.1 | - | 5 | 1058 | |||
AK293950.1 | - | 9 | 1042 | |||
AK295896.1 | - | 21 | 2701 | |||
BC014159.2 | - | 5 | 460 | |||
BC146787.1 | - | 28 | 3977 | 9007246 | ||
BX648137.1 | - | 6 | 2358 | |||
BX648464.1 | - | 19 | 3113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
