siRNA Details
Gene Symbol: DIXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278542.1 | NP_001265471.1 | 6 | 1458 | ||
GenBank mRNA | BC048294.1 | - | 5 | 1171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
