siRNA Details
Gene Symbol: DIXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037954.3 | NP_001033043.1 | 3 | 593 | ||
NM_001278542.1 | NP_001265471.1 | 4 | 573 | |||
XM_017018466.1 | XP_016873955.1 | 4 | 553 | |||
XM_017018467.1 | XP_016873956.1 | 4 | 421 | |||
GenBank mRNA | BC035509.2 | - | 3 | 409 | ||
BC048294.1 | - | 3 | 286 | |||
BC064479.1 | - | 3 | 409 | |||
BC128600.1 | - | 3 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
