siRNA Details
Gene Symbol: LPGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320808.1 | NP_001307737.1 | 5 | 1349 | ||
NM_014873.2 | NP_055688.1 | 5 | 760 | |||
XM_005273362.3 | XP_005273419.1 | 6 | 908 | |||
XM_011510228.2 | XP_011508530.1 | 6 | 797 | |||
XM_011510229.2 | XP_011508531.1 | 5 | 680 | |||
XM_017003004.1 | XP_016858493.1 | 4 | 1912 | |||
GenBank mRNA | AY561706.1 | - | 4 | 533 | ||
BC034621.1 | - | 5 | 1350 | 4839400 | ||
D86960.1 | - | 5 | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
