siRNA Details
Gene Symbol: SDHAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003265.3 | 4, 5 | 521 | |||
GenBank mRNA | AK074764.1 | - | 4, 5 | 467 | ||
BC053551.1 | - | 4, 5 | 512 | |||
BC062438.1 | - | 4, 5 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
