siRNA Details
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 2, 3 | 336 | ||
NM_001281711.1 | NP_001268640.1 | 2, 3 | 297 | |||
NM_001281712.1 | NP_001268641.1 | 1, 2 | 116 | |||
NM_015341.4 | NP_056156.2 | 2, 3 | 369 | |||
XM_005263908.3 | XP_005263965.1 | 2, 3 | 309 | |||
XM_006712388.3 | XP_006712451.1 | 2, 3 | 309 | |||
GenBank mRNA | AK291053.1 | - | 2, 3 | 310 | ||
AK299248.1 | - | 2, 3 | 238 | |||
AK303725.1 | - | 1, 2 | 61 | |||
AK308050.1 | - | 2, 3 | 276 | |||
AK310649.1 | - | 2, 3 | 237 | |||
BC024211.2 | - | 2, 3 | 323 | 3504782 | ||
BI559873.1 | - | 2, 3 | 357 | |||
BI560881.1 | - | 2, 3 | 374 | |||
D38553.1 | - | 2, 3 | 299 |
