siRNA Details
Gene Symbol: CCDC88C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080414.3 | NP_001073883.2 | 28 | 4877 | ||
XM_011536796.2 | XP_011535098.1 | 28 | 4831 | |||
XM_017021336.1 | XP_016876825.1 | 13 | 1940 | |||
GenBank mRNA | BC035914.1 | - | 4 | 589 | ||
BX248302.1 | - | 4 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
