siRNA Details
Gene Symbol: SLX4IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009608.2 | NP_001009608.1 | 6 | 555 | ||
XM_017027647.1 | XP_016883136.1 | 5 | 4057 | |||
XM_017027648.1 | XP_016883137.1 | 7 | 1508 | |||
XM_017027649.1 | XP_016883138.1 | 7 | 701 | |||
XM_017027650.1 | XP_016883139.1 | 7 | 1468 | |||
XM_017027651.1 | XP_016883140.1 | 4 | 506 | |||
XM_017027652.1 | XP_016883141.1 | 6 | 1457 | |||
XM_017027653.1 | XP_016883142.1 | 5 | 4065 | |||
GenBank mRNA | BC020787.1 | - | 6 | 555 | ||
BC026094.1 | - | 6 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
