siRNA Details
Gene Symbol: SLX4IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009608.2 | NP_001009608.1 | 8 | 1330 | ||
XM_017027647.1 | XP_016883136.1 | 7 | 4832 | |||
XM_017027648.1 | XP_016883137.1 | 9 | 2283 | |||
XM_017027649.1 | XP_016883138.1 | 9 | 1476 | |||
XM_017027650.1 | XP_016883139.1 | 9 | 2243 | |||
XM_017027651.1 | XP_016883140.1 | 6 | 1281 | |||
XM_017027652.1 | XP_016883141.1 | 8 | 2232 | |||
GenBank mRNA | BC020787.1 | - | 8 | 1330 | ||
BC026094.1 | - | 8 | 1334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
