siRNA Details
Gene Symbol: SLX4IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009608.2 | NP_001009608.1 | 8 | 911 | ||
XM_017027647.1 | XP_016883136.1 | 7 | 4413 | |||
XM_017027648.1 | XP_016883137.1 | 9 | 1864 | |||
XM_017027649.1 | XP_016883138.1 | 9 | 1057 | |||
XM_017027650.1 | XP_016883139.1 | 9 | 1824 | |||
XM_017027651.1 | XP_016883140.1 | 6 | 862 | |||
XM_017027652.1 | XP_016883141.1 | 8 | 1813 | |||
GenBank mRNA | BC020787.1 | - | 8 | 911 | ||
BC026094.1 | - | 8 | 915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
