siRNA Details
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080448.2 | NP_001073917.2 | 10 | 2214 | ||
NM_001278300.1 | NP_001265229.1 | 7 | 626 | |||
NM_173655.3 | NP_775926.1 | 8 | 742 | |||
XM_006713592.3 | XP_006713655.1 | 11 | 2262 | |||
XM_017006210.1 | XP_016861699.1 | 11 | 2262 | |||
XM_017006211.1 | XP_016861700.1 | 12 | 2287 | |||
XM_017006212.1 | XP_016861701.1 | 11 | 1956 | |||
XM_017006213.1 | XP_016861702.1 | 11 | 2314 | |||
XM_017006214.1 | XP_016861703.1 | 10 | 1582 | |||
XM_017006215.1 | XP_016861704.1 | 6 | 611 | |||
XM_017006216.1 | XP_016861705.1 | 5 | 561 | |||
XM_017006217.1 | XP_016861706.1 | 7 | 658 | |||
XM_017006218.1 | XP_016861707.1 | 5 | 461 | |||
XM_017006220.1 | XP_016861709.1 | 7 | 626 | |||
XM_017006221.1 | XP_016861710.1 | 5 | 572 | |||
XM_017006222.1 | XP_016861711.1 | 6 | 620 | |||
XM_017006223.1 | XP_016861712.1 | 7 | 626 | |||
GenBank mRNA | AK302040.1 | - | 7 | 609 | ||
AK302080.1 | - | 6 | 516 | |||
AK310249.1 | - | 6 | 545 | |||
AL133666.1 | - | 7 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
