siRNA Details
Gene Symbol: HAUS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011699.1 | NP_001011699.1 | 3 | 223 | ||
NM_033417.1 | NP_219485.1 | 3 | 223 | |||
XM_005260154.4 | XP_005260211.1 | 3 | 138 | |||
GenBank mRNA | AK295994.1 | - | 3 | 295 | ||
AY211919.1 | - | 3 | 125 | |||
BI829542.1 | - | 3 | 223 | |||
BM563712.1 | - | 3 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
