siRNA Details
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 7 | 715 | ||
NM_001162501.1 | NP_001155973.1 | 4 | 516 | |||
NM_015088.2 | NP_055903.2 | 4 | 516 | |||
GenBank mRNA | AB029016.2 | - | 4 | 499 | ||
AK294519.1 | - | 4 | 507 | |||
AK296590.1 | - | 4 | 509 | |||
BC028626.2 | - | 7 | 648 | 4826787 | ||
BX952326.1 | - | 4 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
