siRNA Details
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 18 | 2195 | ||
NM_001162501.1 | NP_001155973.1 | 17 | 4516 | |||
NM_015088.2 | NP_055903.2 | 15 | 4186 | |||
GenBank mRNA | AB029016.2 | - | 15 | 4166 | ||
AK294519.1 | - | 17 | 4507 | |||
AK296590.1 | - | 8 | 1235 | |||
AK304087.1 | - | 12 | 1671 | |||
BC028626.2 | - | 18 | 2128 | 4826787 | ||
BX647630.1 | - | 12 | 3388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
